Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIAH2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB317620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317620 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317620 Supplier Novus Biologicals Supplier No. NBP299033PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIAH2 Polyclonal antibody specifically detects SIAH2 in Human samples. It is validated for Western Blot

Specifications

Antigen SIAH2
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias E3 ubiquitin-protein ligase SIAH2, EC 6.3.2, EC 6.3.2.-, hSiah2, seven in absentia (Drosophila) homolog 2, Seven in absentia homolog 2, seven in absentia homolog 2 (Drosophila), siah-2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SIAH2.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6478
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.