Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIDT2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB190444 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB190444 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB190444 Supplier Novus Biologicals Supplier No. NBP282009AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIDT2 Polyclonal antibody specifically detects SIDT2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen SIDT2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias DKFZp686L17253, FLJ90656, SID1 transmembrane family member 2, SID1 transmembrane family, member 2, UNQ685/PRO1325
Host Species Rabbit
Immunogen Rabbit polyclonal SIDT2 antibody was raised against an 18 amino acid peptide near the amino terminus of human SIDT2. The immunogen is located within the first 50 amino acids of SIDT2. Amino Acid Squence: PKNVSQKDAEFERTYVDE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 51092
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.