Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIDT2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017320 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017320 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017320 Supplier Novus Biologicals Supplier No. NBP282009
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIDT2 Polyclonal specifically detects SIDT2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SIDT2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 μg/mL, ELISA
Formulation PBS
Gene Alias DKFZp686L17253, FLJ90656, SID1 transmembrane family member 2, SID1 transmembrane family, member 2, UNQ685/PRO1325
Gene Symbols SIDT2
Host Species Rabbit
Immunogen Rabbit polyclonal SIDT2 antibody was raised against an 18 amino acid peptide near the amino terminus of human SIDT2. The immunogen is located within the first 50 amino acids of SIDT2. Amino Acid Squence: PKNVSQKDAEFERTYVDE
Molecular Weight of Antigen 92 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51092
Test Specificity Multiple isoforms of SIDT2 are known to exist; this antibody is predicted to not cross-react with other members of the SID1 transmembrane family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.