Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIGIRR Antibody (010), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB069454 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB069454 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB069454 Supplier Novus Biologicals Supplier No. NBP290067AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SIGIRR Monoclonal specifically detects SIGIRR in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SIGIRR
Applications Flow Cytometry
Classification Monoclonal
Clone 10
Conjugate Alexa Fluor 350
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias single Ig IL-1-related receptor, Single Ig IL-1R-related molecule, single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, single immunoglobulin domain IL1R1 related, Single immunoglobulin domain-containing IL1R-related protein, TIR8MGC110992, Toll/interleukin-1 receptor 8
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SIGIRR (Q6IA17-1; Met1-His118).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59307
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.