Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIGIRR Antibody (016), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB069495 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB069495 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB069495 Supplier Novus Biologicals Supplier No. NBP290068V
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SIGIRR Monoclonal specifically detects SIGIRR in Human samples. It is validated for ELISA.

Specifications

Antigen SIGIRR
Applications ELISA
Classification Monoclonal
Clone 16
Conjugate DyLight 405
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias single Ig IL-1-related receptor, Single Ig IL-1R-related molecule, single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, single immunoglobulin domain IL1R1 related, Single immunoglobulin domain-containing IL1R-related protein, TIR8MGC110992, Toll/interleukin-1 receptor 8
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SIGIRR (Q6IA17-1; Met1-His118).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59307
Test Specificity No cross-reactivity in ELISA with: Mouse SIGIRR Human IL1R1 Human IL1R2 Human IL1R3 Human IL1R7 Human IL1R8 Human IL1R9 Human IL18R1
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.