Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-1/CD169 Antibody (7-239) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP237729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237729 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237729 Supplier Novus Biologicals Supplier No. NBP237729
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen Siglec-1/CD169
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay, CyTOF
Classification Monoclonal
Clone 7-239
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1 to 4 μg/mL, Immunohistochemistry, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready
Formulation PBS (pH 7.4) with 15mM Sodium Azide
Gene Alias CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin
Gene Symbols SIGLEC1
Host Species Mouse
Immunogen Human rhinovirus 14-infected monocyte-derived dendritic cells.
Molecular Weight of Antigen 210 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6614
Test Specificity The mouse monoclonal antibody 7-239 recognizes CD169 (sialoadhesin, Siglec-1), a 210 kDa type I transmembrane glycoprotein expressed on macrophages and dendritic cells.
Reconstitution Protein A purified
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.