Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-1/CD169 Antibody (908102), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB368358 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368358 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368358 Supplier Novus Biologicals Supplier No. FAB5197L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Siglec-1/CD169
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 908102
Conjugate DyLight 550
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-1/CD169, Ser20-Gln1641, Accession # Q9BZZ2
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6614
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.