Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-1/CD169 Antibody (ED3), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB10065301Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065301Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065301Z Supplier Novus Biologicals Supplier No. NB10065301AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.

Specifications

Antigen Siglec-1/CD169
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone ED3
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin
Gene Symbols SIGLEC1
Host Species Mouse
Immunogen Rat Spleen cell homogenate
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6614
Test Specificity NB100-65301 recognizes the rat CD169 cell surface antigen, a 185kD molecule expressed by macrophages, predominately confined to lymphoid organs only. Monocytes and granulocytes are negative. No other cell types are positive. The most conspicious property of ED3 is it stains marginal zone macrophages and marginal metallophils in the spleen very strongly. Furthermore, macrophages in (auto-immune) diseased tissues express the ED3 antigen. In healthy tissue no expression occurs. CD169 is a receptor for glycoconjugates containing sialic acid.
Reconstitution Protein A purified
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.