Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-1/CD169 Antibody (HSn 7D2), HRP, Novus Biologicals™
SDP

Catalog No. NB600534H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600534H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600534H Supplier Novus Biologicals Supplier No. NB600534H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Human, Mouse, Porcine, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Siglec-1/CD169
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HSn 7D2
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS with No Preservative
Gene Alias CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin
Gene Symbols SIGLEC1
Host Species Mouse
Immunogen Full-length purified human Sialoadhesin (d1-4)Fc protein. [Swiss-Prot# Q9BZZ2]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6614
Target Species Human, Mouse, Pig, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.