Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-10 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15924720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15924720 20 μL
NBP159247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15924720 Supplier Novus Biologicals Supplier No. NBP15924720UL

Rabbit Polyclonal Antibody

Siglec-10 Polyclonal specifically detects Siglec-10 in Human samples. It is validated for Western Blot.

Specifications

Antigen Siglec-10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96LC7
Gene Alias MGC126774, PRO940, sialic acid binding Ig-like lectin 10, sialic acid-binding Ig-like lectin 10, SIGLEC-10, siglec-like gene 2, Siglec-like protein 2, SLG2sialic acid binding Ig-like lectin 10 Ig-like lectin 7
Gene Symbols SIGLEC10
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SIGLEC10(sialic acid binding Ig-like lectin 10) The peptide sequence was selected from the middle region of SIGLEC10. Peptide sequence CHVDFSRKGVSAQRTVRLRVAYAPRDLVISISRDNTPALEPQPQGNVPYL.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89790
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.