Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-11 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB214823 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB214823 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB214823 Supplier Novus Biologicals Supplier No. NBP177025FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Siglec-11 Polyclonal antibody specifically detects Siglec-11 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Siglec-11
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias sialic acid binding Ig-like lectin 11, sialic acid-binding Ig-like lectin 11, Sialic acid-binding lectin 11, siglec-11
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human Siglec11. The immunogen is located within the last 50 amino acids of Siglec11. Amino Acid Squence: WEPADQEAPSTTEYSEIK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 114132
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.