Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-14 Antibody (2457E), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347517 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347517 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347517 Supplier Novus Biologicals Supplier No. FAB4905MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Siglec-14 Monoclonal antibody specifically detects Siglec-14 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Siglec-14
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2457E
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AC018755.2, sialic acid binding Ig-like lectin 14
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human Siglec-14, Lys18-Leu358, Accession # Q08ET2
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100049587
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.