Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-15 Antibody (1026113), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB310530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310530 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310530 Supplier Novus Biologicals Supplier No. FAB9227APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-15 Monoclonal specifically detects Siglec-15 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-15
Applications Flow Cytometry
Classification Monoclonal
Clone 1026113
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD33 antigen-like 3SIGLEC-15, CD33 molecule-like 3, CD33L3, HsT1361, sialic acid binding Ig-like lectin 15, sialic acid-binding Ig-like lectin 15, Siglec-15
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human Siglec-15, Phe20-Thr263, Accession # Q6ZMC9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 284266
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.