Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-15 Antibody (1026113), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB345810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345810 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345810 Supplier Novus Biologicals Supplier No. FAB9227MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-15 Monoclonal specifically detects Siglec-15 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Siglec-15
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 1026113
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD33 antigen-like 3SIGLEC-15, CD33 molecule-like 3, CD33L3, HsT1361, sialic acid binding Ig-like lectin 15, sialic acid-binding Ig-like lectin 15, Siglec-15
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human Siglec-15, Phe20-Thr263, Accession # Q6ZMC9
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 284266
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.