Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-15 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB099022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB099022 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB099022 Supplier Novus Biologicals Supplier No. NBP241162JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Siglec-15 Polyclonal specifically detects Siglec-15 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Siglec-15
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50 mM Sodium Borate
Gene Alias CD33 antigen-like 3SIGLEC-15, CD33 molecule-like 3, CD33L3, HsT1361, sialic acid binding Ig-like lectin 15, sialic acid-binding Ig-like lectin 15, Siglec-15
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human SIGLEC15. The immunogen is located within amino acids 50 - 100 of SIGLEC15. Amino Acid Squence: TFTHPHRHYDGPLT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284266
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.