Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-16 Antibody (706030) [Alexa Fluor™ 532], Novus Biologicals™
SDP

Catalog No. NB375671 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375671 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375671 Supplier Novus Biologicals Supplier No. FAB68192X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-16 Monoclonal antibody specifically detects Siglec-16 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen Siglec-16
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 706030
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Siglec-P16, SIGLECP16
Host Species Mouse
Immunogen Human Siglec-16 peptide, Accession # A6NMB1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400709
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.