Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-2/CD22 Antibody (hL22 (Epratuzumab)) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB001491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001491 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001491 Supplier Novus Biologicals Supplier No. NBP2751880.025MG
Add to Cart
Add to Cart

Human Monoclonal Antibody

Siglec-2/CD22 Monoclonal specifically detects Siglec-2/CD22 in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry, Immunoprecipitation, Block/Neutralize.

Specifications

Antigen Siglec-2/CD22
Applications Flow Cytometry, Immunoprecipitation, Blocking Assay, Neutralization
Classification Monoclonal
Clone hL22 (Epratuzumab)
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Block/Neutralize
Formulation PBS
Gene Alias B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14
Gene Symbols CD22
Host Species Human
Immunogen This antibody was prepared by the humanization of LL2 (EPB-2), a murine anti-CD22 IgG2a raised against Raji Burkitt lymphoma cells. Murine sequences comprise 5-10% of the molecule, with the remainder being human framework sequences, which greatly reduces the potential for immunogenicity (Traczewski, 2010).
Quantity 0.025 mg
Gene ID (Entrez) 933
Test Specificity This antibody is specific for the 3rd Ig-like domain of human CD22 (epitope B), a cell surface glycoprotein present on mature B-cells and on many types of malignant B-cells.
Target Species Human, Cynomolgus Monkey, Rhesus Macaque
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.