Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-3/CD33 Antibody (2337A) [DyLight 650], Novus Biologicals™
SDP

Catalog No. NB400912 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400912 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400912 Supplier Novus Biologicals Supplier No. FAB11373W
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Siglec-3/CD33
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2337A
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human Siglec-3/CD33, Asp18-His259, Accession # P20138
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 945
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.