Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-3/CD33 Antibody (HIM3-4), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB500508IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500508IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500508IR Supplier Novus Biologicals Supplier No. NB500508IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-3/CD33
Applications Flow Cytometry
Classification Monoclonal
Clone HIM3-4
Conjugate DyLight 755
Dilution Flow Cytometry
Gene Alias CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Gene Symbols CD33
Host Species Mouse
Immunogen NFMY-9s human cell line
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 945
Test Specificity The antibody HIM3-4 reacts with CD33, a 67 kDa type I transmembrane glycoprotein (immunoglobulin superfamily) expressed on myeloid progenitors, monocytes, granulocytes, dendritic cells and mast cells; it is absent on platelets, lymphocytes, erythrocytes and hematopoietic stem cells. HLDA V; WS Code M MA112 HLDA VI; WS Code M MA47
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.