Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-3/CD33 Antibody (hP67.6 (Gemtuzumab)) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. N27590602M Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N27590602M 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N27590602M Supplier Novus Biologicals Supplier No. NBP2759060.2MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.

Specifications

Antigen Siglec-3/CD33
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone Gemtuzumab
Conjugate Unconjugated
Dilution Flow Cytometry, Immunofluorescence
Formulation PBS with 0.02% Proclin 300
Gene Alias CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Gene Symbols CD33
Host Species Rabbit
Immunogen CD33 is a transmembrane receptor expressed on predominantly on cells of the myeloid lineage, but it can also be found on some lymphoid cells.
Quantity 0.2 mg
Primary or Secondary Primary
Gene ID (Entrez) 945
Test Specificity Recognises human CD33.
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.