Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-3/CD33 Antibody, Novus Biologicals™
SDP

Catalog No. NBP229619SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229619SS 0.025 mg
NBP229619 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229619SS Supplier Novus Biologicals Supplier No. NBP229619SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Siglec-3/CD33 Polyclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Siglec-3/CD33
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry reported in scientific literature (PMID 27535972), ELISA reported in scientific literature (PMID 27535972), Immunohistochemistry 5 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P20138
Gene Alias CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Gene Symbols CD33
Host Species Rabbit
Immunogen Partial recombinant protein made to an internal portion of human CD33 protein (between residues 30-200) [UniProt P20138]
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 945
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.