Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-5/CD170 Antibody (110), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB068562 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB068562 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB068562 Supplier Novus Biologicals Supplier No. NBP289986AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Siglec-5/CD170 Monoclonal specifically detects Siglec-5/CD170 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-5/CD170
Applications Flow Cytometry
Classification Monoclonal
Clone 110
Conjugate Alexa Fluor 532
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD170, CD170 antigen, CD33 antigen-like 2, CD33L2OB binding protein-2, OB-BP2OB-binding protein 2, OBBP2sialic acid-binding Ig-like lectin 5, Obesity-binding protein 2, sialic acid binding Ig-like lectin 5, sialic acid-binding immunoglobulin-like lectin 5, SIGLEC-5
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Siglec-5/CD170 (O15389; Met1-Thr 434).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 8778
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.