Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-6/CD327 Antibody (767329R), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB363947 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363947 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363947 Supplier Novus Biologicals Supplier No. FAB2859RI
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-6/CD327 Monoclonal antibody specifically detects Siglec-6/CD327 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Siglec-6/CD327
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 767329R
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD327 antigen, CD33 antigen-like 1, CD33L1OBBP1CD327, CD33L2, CD33LCDW327, CDw327, OB-BP1, Obesity-binding protein 1, sialic acid binding Ig-like lectin 6, sialic acid-binding Ig-like lectin 6, Siglec-6
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Siglec-6/CD327, Gln27-Ser331, Accession # O43699
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 946
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.