Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-7/CD328 Antibody (194212), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB297335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297335 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297335 Supplier Novus Biologicals Supplier No. FAB1138L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-7/CD328 Monoclonal specifically detects Siglec-7/CD328 in Human samples. It is validated for Western Blot.

Specifications

Antigen Siglec-7/CD328
Applications Western Blot
Classification Monoclonal
Clone 194212
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec-7
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-7/CD328, Gln19-Gly357 (predicted), Accession # Q9Y286
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 27036
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.