Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-7/CD328 Antibody (OTI1A1), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP274187A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274187A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274187A70 Supplier Novus Biologicals Supplier No. NBP274187AF750

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Siglec-7/CD328 Monoclonal specifically detects Siglec-7/CD328 in Human samples. It is validated for Western Blot.

Specifications

Antigen Siglec-7/CD328
Applications Western Blot
Classification Monoclonal
Clone OTI1A1
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec-7
Gene Symbols SIGLEC7
Host Species Mouse
Immunogen Full length recombinant protein of human Siglec-7/CD328 (NP_057627) produced in HEK293T cells.
Quantity 0.1 mL
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 27036
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.