Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-8 Antibody (837535) [DyLight 488], Novus Biologicals™
SDP

Catalog No. NB373818 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373818 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373818 Supplier Novus Biologicals Supplier No. FAB7975K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-8 Monoclonal specifically detects Siglec-8 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Siglec-8
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 837535
Conjugate DyLight 488
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD329 antigen, CDw329, MGC59785, SAF2SAF-2, sialic acid binding Ig-like lectin 8, Sialoadhesin family member 2, Siglec-8, SIGLEC8Lsialic acid-binding Ig-like lectin 8
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-8, Met17-Ala363, Accession # Q9NYZ4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27181
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.