Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-9 Antibody (MM0552-6K12) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018948 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018948 0.025 mg
NBP211905 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018948 Supplier Novus Biologicals Supplier No. NBP2119050.025MG

Mouse Monoclonal Antibody

Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Western Blot, Block/Neutralize.

Specifications

Antigen Siglec-9
Applications Western Blot, Blocking Assay
Classification Monoclonal
Clone MM0552-6K12
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Blocking/Neutralizing 50-200 molar excess
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q9Y336
Gene Alias CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9
Gene Symbols SIGLEC9
Host Species Mouse
Immunogen Extracellular domain of recombinant human Siglec-9.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27180
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.