Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-9 Antibody (MM0552-6K12), PE, Novus Biologicals™
SDP

Catalog No. NB361587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361587 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361587 Supplier Novus Biologicals Supplier No. NBP211905PE

Mouse Monoclonal Antibody

Siglec-9 Monoclonal antibody specifically detects Siglec-9 in Human samples. It is validated for Western Blot, Neutralization

Specifications

Antigen Siglec-9
Applications Western Blot, Neutralization
Classification Monoclonal
Clone MM0552-6K12
Conjugate PE
Dilution Western Blot, Block/Neutralize
Formulation PBS
Gene Alias CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9
Host Species Mouse
Immunogen Human recombinant Siglec-9 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 27180
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.