Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-9 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227070SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227070SS 0.025 mg
NBP227070 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227070SS Supplier Novus Biologicals Supplier No. NBP227070SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Siglec-9 Polyclonal specifically detects Siglec-9 in Bovine, Canine, Equine, Monkey, Primate samples. It is validated for Western Blot.

Specifications

Antigen Siglec-9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9Y336
Gene Alias CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9
Gene Symbols SIGLEC9
Host Species Rabbit
Immunogen A portion of amino acids 40-90 of human Siglec9 was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 27180
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, dwarf chimpanzee, common chimpanzee, great ape, 89% in pig and dog, 84% in cow, rhesus monkey, and horse, and 74% homologous in orangutan.
Target Species Bovine, Canine, Equine, Monkey, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.