Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
| Antigen | Siglec-9 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-4ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227070SS
![]() |
Novus Biologicals
NBP227070SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP227070
![]() |
Novus Biologicals
NBP227070 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
Siglec-9 Polyclonal specifically detects Siglec-9 in Bovine, Canine, Equine, Monkey, Primate samples. It is validated for Western Blot.Specifications
| Siglec-9 | |
| Western Blot 2-4ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Canine, Equine, Monkey, Primate | |
| Q9Y336 | |
| 27180 | |
| A portion of amino acids 40-90 of human Siglec9 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9 | |
| SIGLEC9 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, dwarf chimpanzee, common chimpanzee, great ape, 89% in pig and dog, 84% in cow, rhesus monkey, and horse, and 74% homologous in orangutan. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title