Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-9 Antibody (OTI1D9), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP274189V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274189V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274189V Supplier Novus Biologicals Supplier No. NBP274189V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-9 Monoclonal specifically detects Siglec-9 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Siglec-9
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1D9
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9
Gene Symbols SIGLEC9
Host Species Mouse
Immunogen Full length recombinant protein of human Siglec-9 (NP_055256) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 27180.0
Target Species Human, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.