Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-9 Antibody (MM0552-6K12) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
| Antigen | Siglec-9 |
|---|---|
| Clone | MM0552-6K12 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:500-1:1000, Blocking/Neutralizing 50-200 molar excess |
| Applications | Western Blot, Blocking Assay |
Description
Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Western Blot, Block/Neutralize.Specifications
| Siglec-9 | |
| LYOPH | |
| Western Blot, Blocking Assay | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9 | |
| SIGLEC9 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0552-6K12 | |
| Western Blot 1:500-1:1000, Blocking/Neutralizing 50-200 molar excess | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9Y336 | |
| 27180 | |
| IgG2 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title