Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-H Antibody (23M14A10), PE, Novus Biologicals™
SDP

Catalog No. NBP227061PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227061PE 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227061PE Supplier Novus Biologicals Supplier No. NBP227061PE
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Siglec-H Monoclonal specifically detects Siglec-H in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-H
Applications Flow Cytometry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone 23M14A10
Concentration 1.0 mg/mL
Conjugate PE
Dilution Flow Cytometry 1ug/10^6cells
Formulation PBS with 0.05% Sodium Azide
Gene Alias 6430529G09Rik, Siglec-H, Siglech sialic acid binding Ig-like lectin H
Gene Symbols Siglech
Host Species Rat
Immunogen SkRSHTkRVERPATHMDQA (aa286-304)
Purification Method Protein G purified
Quantity 0.05 mL
Primary or Secondary Primary
Gene ID (Entrez) 233274
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.