Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-H Antibody (440c), Novus Biologicals™
SDP

Catalog No. NB119860 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119860 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119860 Supplier Novus Biologicals Supplier No. NBP311235
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Siglec-H Monoclonal specifically detects Siglec-H in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional.

Specifications

Antigen Siglec-H
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay
Classification Monoclonal
Clone 440c
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias 6430529G09Rik, Siglec-H, Siglech sialic acid binding Ig-like lectin H
Host Species Rat
Immunogen The monoclonal antibody 440c reacts with Siglec-H, a cell-surface receptor molecule selectively expressed on murine natural interferon producing cells (IPC), also called mouse plasmacytoid dendritic cells (pDC).
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233274
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.