Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-H, Rat anti-Mouse, Clone: 551, Millipore Sigma™
Rat Monoclonal Antibody
$192.34 - $482.80
Specifications
Antigen | Siglec-H |
---|---|
Clone | 551 |
Dilution | Immunohistochemistry Analysis: A representative lot detected Siglec-H in Immunohistochemistry applications (Yun, T.J., et. al. (2016). Cell Metab. 23(5):852-66).Flow Cytometry Analysis: A representative lot detected Siglec-H in Flow Cytometry applications (Yun, T.J., et. al. (2016). Cell Metab. 23(5):852-66). |
Applications | Flow Cytometry, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF204525
|
MilliporeSigma
MABF204525UG |
25 μg |
Each for $192.34
|
|
|||||
MABF2045100
|
MilliporeSigma
MABF2045100UG |
100 μg |
Each for $482.80
|
|
|||||
Description
Anti-Siglec-H, clone 551, Cat. No. MABF2045, is a rat monoclonal antibody that detects Sialic acid binding Ig-like lectin H and has been tested for use in Flow Cytometry and Immunohistochemistry.
Siglec-H (UniProt: Q3Y597; also known as Sialic acid binding Ig-like lectin H) is encoded by the Siglech gene (Gene ID: 233274) in murine species. Siglecs are sialic acid-binding immunoglobulin-like lectins that are expressed mostly in the immune system and may be involved in both adhesive and signaling functions. Siglec-H is a type I, CD33-related siglec-like transmembrane glycoprotein that is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off in the mature form. The mature form has to ig-like domains (aa 32-141 and 151-234) in its extracellular region. Unlike other CD33-related siglecs, it lacks tyrosine-based signaling motifs in its cytoplasmic tail. Siglec-H is shown to be expressed on plasmaocytoid dendritic cells (pDCs) and can serve as a reliable marker for these cells. Siglec-H is also reported to serve as an endocytic receptor expressed on murine pDCs. Action of Siglec-H depends on a 12 kDa DNAX-activating protein (DAP). (Ref.: Zhang, J., et al. (2006). Blood 107(9); 3600-3608; Yun, TJ., et al., (2016), Cell Metabolism 23(5); 852-866).Specifications
Siglec-H | |
Immunohistochemistry Analysis: A representative lot detected Siglec-H in Immunohistochemistry applications (Yun, T.J., et. al. (2016). Cell Metab. 23(5):852-66).Flow Cytometry Analysis: A representative lot detected Siglec-H in Flow Cytometry applications (Yun, T.J., et. al. (2016). Cell Metab. 23(5):852-66). | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
Sialic acid binding Ig-like lectin H;Siglech | |
Siglech;Siglec-H | |
IgG1 κ | |
Protein G purified | |
Clone 551 is a rat monoclonal antibody that detects murine Siglec-H. It targets an epitope with in the extracellular domain. |
551 | |
Flow Cytometry, Immunohistochemistry | |
Unconjugated | |
Rat | |
Inflammation & Immunology | |
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_848821.2 | |
Bone marrow-derived Type 1 interferon-producing cells. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title