Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC14 Polyclonal Antibody

Rabbit Polyclonal Antibody

$237.00 - $539.50

Specifications

Antigen SIGLEC14
Concentration 0.5 mg/mL
Content And Storage -20°C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119412
View Documents
Invitrogen™
PA5119412
200 μL
Each for $539.50
Only null left
Add to Cart
 
PIPA5121610
View Documents
Invitrogen™
PA5121610
20 μL
Each for $237.00
Only null left
Add to Cart
 
Description

Description

SIGLEC14 Polyclonal Antibody for Western Blot, IHC (P)

Putative adhesion molecule. Sialic acid-binding paired receptor which may activate associated receptors. Interacts with TYROBP. Mainly expressed in hematopoietic tissues including bone marrow, spleen and fetal liver. Siglec-14 is predominantly expressed in hematopoietic tissues but is also found in lung and testis.
Specifications

Specifications

SIGLEC14
-20°C
Polyclonal
Liquid
IgG
Human
SIGLEC14
LLNLR-470E3.1; sialic acid binding Ig like lectin 14; sialic acid binding Ig-like lectin 14; sialic acid-binding Ig-like lectin 14; SIG14; Siglec; SIGLEC14; Siglec-14; UNQ294; UNQ294/PRO333
SIGLEC14
Primary
Antigen Affinity Chromatography
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300
Q08ET2
100049587
Recombinant protein Sialic Acid Binding Ig Like Lectin 14. The antigen corresponds to amino acid range 24-137 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.