Learn More
Invitrogen™ SIGLEC15 Polyclonal Antibody

Description
The antibody detects endogenous levels of total SIGLEC15 protein.
Specifications
Specifications
| Antigen | SIGLEC15 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | SIGLEC15 |
| Gene Accession No. | Q6ZMC9 |
| Gene Alias | CD33 antigen-like 3; CD33 molecule-like 3; Cd33l3; EG620235; HsT1361; RGD1565948; sialic acid binding Ig like lectin 15; sialic acid binding Ig-like lectin 15; sialic acid-binding Ig-like lectin 15; Siglec; Siglec 15; SIGLEC15; Siglec-15; SIGLEC-I |
| Gene Symbols | SIGLEC15 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.