Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma-1 R/OPRS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP182479
Description
Sigma-1 R/OPRS1 Polyclonal antibody specifically detects Sigma-1 R/OPRS1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Sigma-1 R/OPRS1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
SIGMAR1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:FVFQREEIAQLARQYAGLDHELAFSRLIVELRRLHPGHVLPDEELQWVFVNAGG | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.1mg/mL | |
Western Blot 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:10-1:20 | |
Aging-associated gene 8 protein, FLJ25585, hSigmaR1, OPRS1, SIG-1R, sigma 1, Sigma 1-type opioid receptor, sigma non-opioid intracellular receptor 1, sigma1 receptor, sigma1R, sigma1-receptor, SR31747 binding protein 1, SR31747-binding protein, SR-BP, SR-BP1, SRBPMGC3851, type I sigma receptor | |
Rabbit | |
Affinity Purified | |
RUO | |
10280 | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction