Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Signal sequence receptor delta Antibody (26G5), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP242656UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242656UV 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242656UV Supplier Novus Biologicals Supplier No. NBP242656UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Signal sequence receptor delta Monoclonal specifically detects Signal sequence receptor delta in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Signal sequence receptor delta
Applications Western Blot, ELISA
Classification Monoclonal
Clone 26G5
Conjugate DyLight 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias signal sequence receptor, delta (translocon-associated protein delta), SSR-delta, translocon-associated protein delta, translocon-associated protein subunit delta, TRAP-delta, TRAPDSignal sequence receptor subunit delta
Gene Symbols SSR4
Host Species Mouse
Immunogen Recombinant human SSR4 (24-144aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6748
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.