Learn More
Signal sequence receptor delta Antibody (26G5), FITC, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Signal sequence receptor delta |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 26G5 |
| Conjugate | FITC |
| Dilution | ELISA |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | signal sequence receptor, delta (translocon-associated protein delta), SSR-delta, translocon-associated protein delta, translocon-associated protein subunit delta, TRAP-delta, TRAPDSignal sequence receptor subunit delta |
| Gene Symbols | SSR4 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.