Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Signal sequence receptor delta Antibody (26G5), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309938 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309938 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309938 Supplier Novus Biologicals Supplier No. NBP242656MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Signal sequence receptor delta Monoclonal antibody specifically detects Signal sequence receptor delta in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Signal sequence receptor delta
Applications Western Blot, ELISA
Classification Monoclonal
Clone 26G5
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias signal sequence receptor, delta (translocon-associated protein delta), SSR-delta, translocon-associated protein delta, translocon-associated protein subunit delta, TRAP-delta, TRAPDSignal sequence receptor subunit delta
Host Species Mouse
Immunogen Recombinant human Signal sequence receptor delta (24-144aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6748
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.