Learn More
SIGNR4/CD209e Antibody (RM0327-9B29), Alexa Fluor™ 532, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | SIGNR4/CD209e |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | RM0327-9B29 |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Host Species | Rat |
| Immunogen | Mouse recombinant SIGNR4/CD209e |
| Purification Method | Protein A or G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.