Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIGNR4/CD209e Antibody (RM0327-9B29), HRP, Novus Biologicals™
SDP

Catalog No. NBP212431H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212431H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212431H Supplier Novus Biologicals Supplier No. NBP212431H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SIGNR4/CD209e Monoclonal specifically detects SIGNR4/CD209e in Mouse samples. It is validated for Western Blot.

Specifications

Antigen SIGNR4/CD209e
Applications Western Blot
Classification Monoclonal
Clone RM0327-9B29
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Host Species Rat
Immunogen Mouse recombinant SIGNR4/CD209e
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.