Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIK1/Snf1lk Antibody, Novus Biologicals™
SDP

Catalog No. NBP238397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP238397 0.1 mL
NB396845 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP238397 Supplier Novus Biologicals Supplier No. NBP238397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIK1/Snf1lk Polyclonal specifically detects SIK1/Snf1lk in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SIK1/Snf1lk
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P57059
Gene Alias EC 2.7.11, EC 2.7.11.1, msk, myocardial SNF1-like kinase, salt-inducible kinase 1, Salt-inducible protein kinase 1, serine/threonine protein kinase, serine/threonine-protein kinase SIK1, Serine/threonine-protein kinase SNF1-like kinase 1, Serine/threonine-protein kinase SNF1LK, SIK, SIK-1, SNF1-like kinase, SNF1LKMSK
Gene Symbols SIK1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: YLLLERLKEYRNAQCARPGPARQPRPRSSDLSGLEVPQEGLSTDPFRPALLCPQPQTLVQSVLQAEMDCELQSSLQWPLFFPVDASCSGVF
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 150094
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.