Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIK3 Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP2411470.025MG
Description
SIK3 Polyclonal specifically detects SIK3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence.Specifications
SIK3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
SIK3 | |
Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SIK3. The immunogen is located within amino acids 1190 - 1240 of SIK3. Amino Acid Squence: CHRPRALQRHHTIQNSDDA | |
0.025 mg | |
Protein Kinase | |
23387 | |
Human, Mouse, Rat | |
IgY |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
EC 2.7.11, EC 2.7.11.1, KIAA0999L19, QSKFLJ12240, Salt-inducible kinase 3, Serine/threonine-protein kinase QSK, serine/threonine-protein kinase SIK3, SIK family kinase 3, SIK-3 | |
Chicken | |
Affinity Purified | |
RUO | |
Primary | |
At least three isoforms of SIK3 are known to exist; this antibody will detect two isoforms. SIK3 antibody is predicted to not cross-react with SIK1 and SIK2. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction