Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIK3 Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
$196.10 - $501.50
Specifications
Antigen | SIK3 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SIK3 Polyclonal specifically detects SIK3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence.Specifications
SIK3 | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.11, EC 2.7.11.1, KIAA0999L19, QSKFLJ12240, Salt-inducible kinase 3, Serine/threonine-protein kinase QSK, serine/threonine-protein kinase SIK3, SIK family kinase 3, SIK-3 | |
SIK3 | |
IgY | |
Affinity Purified |
1 mg/mL | |
Polyclonal | |
Chicken | |
Protein Kinase | |
PBS with 0.02% Sodium Azide | |
23387 | |
Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SIK3. The immunogen is located within amino acids 1190 - 1240 of SIK3. Amino Acid Squence: CHRPRALQRHHTIQNSDDA | |
Primary | |
At least three isoforms of SIK3 are known to exist; this antibody will detect two isoforms. SIK3 antibody is predicted to not cross-react with SIK1 and SIK2. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title