Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tube
Click to view available options
Volume (Metric):
0.6 mL
1.5 mL
2 mL
Description
The importance of low DNA binding microcentrifuge tubes cannot be overstated. These tubes are specifically engineered to minimize the adhesion of DNA to the tube walls, which is crucial in preserving the quantity and quality of DNA samples during various laboratory procedures.
Ensuring that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results.
Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.
Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.

Specifications
Specifications
Autoclavable | Autoclavable |
Color | Natural |
Material | Polypropylene Resin |
Volume (Metric) | 2 mL |
Sterility | Non-sterile |
Graduated | Yes |
Closure Type | Snap Cap |
Shape | Cylindrical |
Fit for Purpose | PCR Ready |
Bottom Shape | Conical |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction