Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica gel 60, 0.019-0.037mm (400-600 mesh)

Catalog No. AA4272930
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg

AA4272918 | CAS: 7631-86-9 | O2Si | 60.08 g/mol

Column chromatography with elevated pressureSilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Physical Form Powder
Color White
Quantity 250 g
Odor Odorless
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Chemical Name or Material Silica gel 60
Hazard Category H373: May cause damage to organs through prolonged or repeated exposure.
EINECSNumber 231-545-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.