Learn More
Silica Gel, Grade 633, 200-425 Mesh, 60A, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Silica Gel, Grade 633, 200-425 Mesh, 60A is a form of silicon dioxide made from sodium silicate. It is granular and porous solid and is a naturally occurring mineral. It is often used as a desiccant to control humidity and avoid spoilage. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 1 |
| Material | Silica Gel |
| Quantity | 1 kg |
| Packaging | Steel Can |
| Pore Volume | 0.70-0.85 CC/G |
| Grade | Ungraded |
| Surface Area | 480 m2/g |
| Chemical Name or Material | Silica Gel |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.