Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicagel, functionalized, (cyclohexylcarbodiimido)propyl, Thermo Scientific™

Catalog No. p-3598478
Click to view available options
Quantity:
1 g

Catalog No. AC371530010

Supplier: Thermo Scientific Chemicals 371530010

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym Cyclohexylcarbodiimide, immobilized on silicagel
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Quantity 1 g
Formula Weight 60.08
Chemical Name or Material Silicagel, Functionalized, (cyclohexylcarbodiimido)propyl, ca. 0.9mmol/g, particle size: 40-63 micron

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.